Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Melon™ Gel IgG Spin Purification Kit PROMO

Catalog No. PI45206PM
Click to view available options
:
Thermo Scientific™ Melon™ Gel IgG Spin Purification Kit

Catalog No. PI45206PM

Supplier: Thermo Scientific™ 45206

Rapidly purify human, mouse and rabbit IgG from serum and other sources with this simpler alternative to traditional Protein A and Protein G methods.

Melon Gel IgG Purification Kits enable fast, easy and gentle purification of antibodies from serum and other sample types. Antibody purification with Melon Gel may be performed with gravity-flow columns, spin columns or chromatography cartridges. Melon Gel IgG Purification saves time compared to classical Protein A or Protein G methods without compromising the quality of the antibody. Compared to antibody affinity purification with Protein A or Protein G, Melon Gel IgG Purification is simpler, faster and provides greater yields and higher purity for a wider variety of antibodies species.

Highlights:

  • Simple, one-step protocol – IgG Purification with Melon Gel Resin does not require the binding, washing, and multiple elution steps required by Protein A and Protein G

  • Rapid purification – antibody purification from serum is four to six times faster than Protein A or G methods

  • High recovery and purity – antibody recovery from serum for many species is greater than 90% with greater than 80% purity

  • Minimal dilution – reduced need to concentrate antibody before storage or use

  • Robust purification – Melon Gel IgG Purification Kits work with a wide range of antibodies including many that do not purify well on Protein A or Protein G

  • Gentle purification – no harsh antibody elution conditions helps retain IgG activity

  • Reusable support – Melon Gel Resin can be used for multiple antibody purifications

  • Available in various formats – spin columns, purification kits and chromatography cartridges

Specifications

Product Type IgG Purification Kit
Product Line Melon™
Column Type Proprietary Resin, Affinity
Format Kit, Microcentrifuge Spin Column
Stationary Phase Proprietary Ligand
Purification Target IgG Antibodies
Content And Storage Sufficient For: IgG purification from approx. 25 x 0.1 mL serum samples (total approx. 25 mg IgG from serum), Melon Gel Support, 3 mL, Melon Gel Purification Buffer, 100 mL, Spin Columns, 0.8 mL, 27 Columns, Microcentrifuge Tubes, 2 mL, 30 Tubes, Store at 4°C
Quantity 3 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.