Learn More
Thermo Scientific™ Melon™ Gel Monoclonal IgG Purification Kit PROMO
Description
The Melon Gel Monoclonal IgG Purification Kit consists of a large package size of Melon Gel Resin and an optimized protocol for easy and mild purification of monoclonal IgG antibodies from hybridoma culture supernatant or ascites fluid. Unlike typical bind-and-elute affinity methods, Melon Gel Purification is based on the binding of non-IgG proteins from a prepared supernatant or ascites sample, thereby allowing the pure IgG to be recovered in the flow-through (non-bound) fraction. Antibodies are recovered ready for use in assays or subsequent conjugation and other modification steps. There is no need to neutralize or desalt to mitigate the effects of harsh elution buffers. Melon Gel Purification saves time compared to classical Protein A or Protein G methods without compromising the quality of the antibody.
Highlights:
Fast – purify antibodies in half the time of required for Protein A or G
Efficient – recovery and purity is often better than Protein A or G purification
Robust – effective for subclasses of IgG that bind poorly to Protein A or G
Gentle – antibodies retain activity because they are not exposed to denaturing conditions
Reusable – support can be regenerated and used for up to 6 purifications
Compatible – amine-free buffer does not require removal or neutralization
Flexible – accommodates a broad range of sample volumes and processing vessels
Includes:
Melon Gel Resin, purification buffer and resin regenerant
Specifications
Specifications
| Product Type | IgG Purification Kit |
| Product Line | Melon |
| Column Type | Proprietary Resin, Affinity |
| Format | Kit, Bottle |
| Stationary Phase | Proprietary Ligand |
| Purification Target | IgG Antibodies |
| Content And Storage | Sufficient For: IgG purification from up to 1 L of cell culture supernatant or up to 200 mL of ascites fluid, Melon Gel Support, 200 mL, Melon Gel Purification Buffer (dry mix to make 100X), 1 L, Melon Gel Regenerant (dry mix to make 5X), 1 L Store at 4°C |
| Quantity | 200 mL |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.