Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Meloxicam Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA175257
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA175257 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA175257 Supplier Invitrogen™ Supplier No. PA175257
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Working Concentration: 0.3125 μg/mL. Sensitivity: 10 ng/mL Meloxicam Sodium salt produces 78% inhibition in a competitive ELISA employing Meloxicam polyclonal antibody.

Meloxicam is an oxicam derivative and a non-steroidal anti-inflammatory drug (NSAID) with anti-inflammatory, antipyretic and analgesic activities. Unlike traditional nonselective NSAIDs, meloxicam preferentially inhibits the activity of cyclo-oxygenase II (COX-II), resulting in a decreased conversion of arachidonic acid into prostaglandin precursors. The resulting decrease in prostaglandin synthesis is responsible for the therapeutic effects of meloxicam. [National Center for Biotechnology Information. PubChem Compound Database; CID=54677470].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Meloxicam
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.6
Gene Alias C14H13N3O4S2; Meloxicamum; Mobic; Mobicox; Movalis
Host Species Sheep
Immunogen Meloxicam-BTG.
Purification Method Purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.