Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Melperone 95.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
25 mg
100 mg
Specifications
Specifications
Melting Point | 80°C |
Boiling Point | 125°C |
Quantity | 25 mg |
Synonym | 4′C-Fluoro-4-(4-methylpiperidino)butyrophenone |
Formula Weight | 263.36 |
Percent Purity | ≥95.0% (GC) |
Physical Form | Crystalline Powder |
Chemical Name or Material | Melperone |
Safety and Handling
TSCA | No |
Recommended Storage | Refrigerator |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction