Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Melphalan-modified DNA Antibody (MP5/73) - BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP2505040.025ML
Description
Melphalan-modified DNA Monoclonal antibody specifically detects Melphalan-modified DNA in Nonspecies specific samples. It is validated for ELISA, ImmunohistochemistrySpecifications
Melphalan-modified DNA | |
Monoclonal | |
1 mg/mL | |
ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG2b |
ELISA, Immunohistochemistry | |
MP5/73 | |
Unconjugated | |
PBS | |
Native DNA reacted with melphalan | |
0.025 mL | |
Cancer | |
Non-species specific | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction