Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Melphalan-modified DNA Antibody (MP5/73), FITC, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP250504F
Description
Melphalan-modified DNA Monoclonal specifically detects Melphalan-modified DNA in Non-species specific samples. It is validated for ELISA, Immunohistochemistry.Specifications
Melphalan | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
Native DNA reacted with melphalan | |
0.1 mL | |
Cancer | |
Other | |
Purified |
ELISA, Immunohistochemistry | |
MP5/73 | |
ELISA, Immunohistochemistry | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction