Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Melting point standard (182-184°C), MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
250 mg
1 g
Description
Cleaning Products, Cosmetics, Flavors and Fragrances, Food and Beverages and Personal Care
Specifications
Specifications
| CAS | 100-09-4 |
| Flash Point | 185°C |
| Physical Form | Neat |
| Linear Formula | CH3 OC6H4 CO2 H |
| MDL Number | MFCD00002542 |
| Melting Point | 182°C to 184°C (±0.3); 182°C to 185°C |
| Synonym | p-Anisic acid; 4-Methoxybenzoic acid; p-Methoxybenzoic acid; Draconic acid |
| Type | Analytical Standard |
| Quantity | 1 g |
| Formula Weight | 152.15 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction