Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Membralin Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. ABN1661MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN1661MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN1661MI Supplier MilliporeSigma Supplier No. ABN1661
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Membralin Antibody is a highly specific rabbit polyclonal antibody that targets Membralin and has been tested in Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Membralin
Applications Western Blot, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols Tmem259,ORF61
Host Species Rabbit
Immunogen A Myc-conjugated linear peptide from the N-terminal region of mouse membralin.
Purification Method Protein A + Protein G
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_001003949
Test Specificity This polyclonal antibody specifically detects membralin in HEK293 cells overexpressing membralin.
Target Species Human, Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.