Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MEMO1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103089
Description
Antibody detects endogenous levels of total MEMO1.
May control cell migration by relaying extracellular chemotactic signals to the microtubule cytoskeleton. Mediator of ERBB2 signaling. The MEMO1-RHOA-DIAPH1 signaling pathway plays an important role in ERBB2-dependent stabilization of microtubules at the cell cortex. It controls the localization of APC and CLASP2 to the cell membrane, via the regulation of GSK3B activity. In turn, membrane-bound APC allows the localization of the MACF1 to the cell membrane, which is required for microtubule capture and stabilization. Is required for breast carcinoma cell migration.
Specifications
MEMO1 | |
Polyclonal | |
Unconjugated | |
MEMO1 | |
0610016J10Rik; C21orf19-like protein; C2orf4; CGI-27; D930048L02Rik; fi04d12; HCV NS5A-transactivated protein 7; Hepatitis C virus NS5A-transactivated protein 7; hypothetical protein LOC327360; mediator of cell motility 1; Mediator of ErbB2-driven cell motility 1; MEMO; MEMO1; Memo-1; Methylation modifier for class I HLA; NS5ATP7; protein memo; protein MEMO1; RGD1309929; Unknown (protein for MGC:127714); wu:fi04d12; zgc:55290 | |
Rabbit | |
Affinity chromatography | |
RUO | |
51072, 7795 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9Y316 | |
MEMO1 | |
A synthesized peptide derived from human MEMO1(Accession Q9Y316), corresponding to amino acid residues M1-A44. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction