Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MEMO1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560043
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560043 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560043 Supplier Invitrogen™ Supplier No. PA560043
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FILGPSHHVP LSRCALSSVD IYRTPLYDLR IDQKIYGELW KTGMFERMSL QTDEDEHSIE MHLPYTAK Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

May control cell migration by relaying extracellular chemotactic signals to the microtubule cytoskeleton. Mediator of ERBB2 signaling. The MEMO1-RHOA-DIAPH1 signaling pathway plays an important role in ERBB2-dependent stabilization of microtubules at the cell cortex. It controls the localization of APC and CLASP2 to the cell membrane, via the regulation of GSK3B activity. In turn, membrane-bound APC allows the localization of the MACF1 to the cell membrane, which is required for microtubule capture and stabilization. Is required for breast carcinoma cell migration.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MEMO1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MEMO1
Gene Accession No. Q9Y316
Gene Alias 0610016J10Rik; C21orf19-like protein; C2orf4; CGI-27; D930048L02Rik; fi04d12; HCV NS5A-transactivated protein 7; Hepatitis C virus NS5A-transactivated protein 7; hypothetical protein LOC327360; mediator of cell motility 1; Mediator of ErbB2-driven cell motility 1; MEMO; MEMO1; Memo-1; Methylation modifier for class I HLA; NS5ATP7; protein memo; protein MEMO1; RGD1309929; Unknown (protein for MGC:127714); wu:fi04d12; zgc:55290
Gene Symbols MEMO1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MEMO1. Recombinant protein control fragment (Product #RP-96171).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51072, 7795
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.