Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEMO1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569400
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569400 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569400 Supplier Thermo Scientific Supplier No. PA569400
Add to Cart
Add to Cart

MEMO1 Polyclonal Antibody

MEMO1 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen MEMO1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MEMO1
Gene Accession No. Q91VH6
Gene Alias 0610016J10Rik; C21orf19-like protein; C2orf4; CGI-27; D930048L02Rik; fi04d12; HCV NS5A-transactivated protein 7; Hepatitis C virus NS5A-transactivated protein 7; hypothetical protein LOC327360; mediator of cell motility 1; Mediator of ErbB2-driven cell motility 1; MEMO; MEMO1; Memo-1; Methylation modifier for class I HLA; NS5ATP7; protein memo; protein MEMO1; RGD1309929; Unknown (protein for MGC:127714); wu:fi04d12; zgc:55290
Gene Symbols MEMO1
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human MEMO1 (aa 191-240).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 76890
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.