Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MEMO1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570243
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 90%; Zebrafish: 100%.
May control cell migration by relaying extracellular chemotactic signals to the microtubule cytoskeleton. Mediator of ERBB2 signaling. The MEMO1-RHOA-DIAPH1 signaling pathway plays an important role in ERBB2-dependent stabilization of microtubules at the cell cortex. It controls the localization of APC and CLASP2 to the cell membrane, via the regulation of GSK3B activity. In turn, membrane-bound APC allows the localization of the MACF1 to the cell membrane, which is required for microtubule capture and stabilization. Is required for breast carcinoma cell migration.
Specifications
MEMO1 | |
Polyclonal | |
Unconjugated | |
MEMO1 | |
0610016J10Rik; C21orf19-like protein; C2orf4; CGI-27; D930048L02Rik; fi04d12; HCV NS5A-transactivated protein 7; Hepatitis C virus NS5A-transactivated protein 7; hypothetical protein LOC327360; mediator of cell motility 1; Mediator of ErbB2-driven cell motility 1; MEMO; MEMO1; Memo-1; Methylation modifier for class I HLA; NS5ATP7; protein memo; protein MEMO1; RGD1309929; Unknown (protein for MGC:127714); wu:fi04d12; zgc:55290 | |
Rabbit | |
Affinity chromatography | |
RUO | |
298787 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q4QQR9 | |
MEMO1 | |
synthetic peptide directed towards the middle region of human MEMO1 (aa 171-220). | |
100 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction