Learn More
Description
Flame resistant/chemical protective lab coat brings together the latest innovations in lab safety to protect wearers against multiple hazards.
- Lightweight and breathable fire resistant/chemical protective lab coat.
- Black lay-down collar for easy recognition as a CP lab coat.
- Splash resistant pass-through for inner pocket access.
- Flame-resistant 4.5 oz. Nomex IIIA fabric prevents clothing ignition during short-duration thermal incidents and emergency exposure to flame.
- Repels small amounts of liquid chemicals that are either dropped or splashed on the fabric, resisting penetration and wicking of an inadvertent, secondary liquid chemical splash.
- Concealed snap front neck closures.
- One double-stitched chest pocket and two double-stitched lower pockets with flaps
- Arc Rating ATPV 5.6 cal/cm2
- Category 1 Protection
- NFPA 2112 Compliant
Specifications
Specifications
| Product Type | Flame-resistant Lab Coat |
| Size | 2X-Large |
| Gender | Men's |
| Material | Aramid Fiber |
| Color | Royal Blue |
| Closure Type | Snaps |
| Cuff Style | Knit |
| Disposable | Reusable |
| Sterility | Non-sterile |
| Resistance | Flame, Chemicals |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.