Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bulwark™ Men's Long Sleeve Grey Flame Resistant Performance T-Shirt

Catalog No. 17204001
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large

Men's Long Sleeve Grey Flame Resistant Performance T-Shirt provides flame-resistant protection in a lightweight blended fabric. The result is unbeatable softness coupled with lasting durability. This casual FR shirt features raglan sleeves as well as rib knit collar and cuffs

  • As part of our CoolTouch™ 2 fabric collection, this long sleeve tee provides flame-resistant protection in a lightweight blended fabric.
  • The result is unbeatable softness coupled with lasting durability.
  • This casual FR shirt features raglan sleeves as well as rib knit collar and cuffs
  • Certifications Level 1: NFPA 2112, NFPA 70E
  • Certifications Level 2: Category 2 Protection, NFPA 2112 - 2018 Compliant
  • Hazard: Flash Fire, Arc Flash
  • Number of Pockets: 1
  • Primary Closure: No Closure
  • Wash Care: Industrial Laundry - Light Soil, Home Wash
  • Fabric: Flame-resistant, 6.5 oz. (220 g/m&superscript_2;) with moisture management finish
  • Blend: 45% Modacrylic / 35% FR Lyocell / 20% Aramid
  • Collar: Rib knit collar
  • Pocket: Single spade chest pocket
  • Features: Heat seal Bulwark™ triangle centered at neck to show wearer has on FR undergarment, Full side seam gusset, Rib knit cuffs and collar, Raglan sleeves for ease of movement, Tagless brand label and warning label
  • Product Collection: Cooltouch 2
  • Packaging: Hang tag
  • Protection: EBT Arc Rating: 8.2
  • Country of Origin: Imported, Made in USA Fabric
  • Fit Details: Regular Fit
  • Gender: Male
  • Materials: Modacrylic/FR Lyocell/Aramid
  • Brand: Bulwark™

Specifications

Size 3X-Large
Gender Men's
Material Modacrylic Blend
Color Gray
Neck Style Hemmed Crew
Sleeve Style Raglan Sleeve
Closure Type Pull On
Disposable Reusable
No. of Pockets 1
Hazard Risk Category Category 2
Arc Flash Rating 8.2
Certifications/Compliance NFPA 2112, NFPA 70E
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.