Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bulwark™ Men's Navy Flame Resistant Flex Knit Button Down Shirt
Click to view available options
Size:
2X-Large
2X-Large (Long)
3X-Large
Large
Large (Long)
Medium
Medium (Long)
Small
X-Large
X-Large (Long)
Description
- Spandex-blend fabric and side seam gussets allows it to stretch freely without coming untucked
- Two reinforced chest pockets with button flaps allows convenient storage
- Banded collar with full underarm gusset for added mobility
- Two chest pockets with flaps and button closure
- Lightweight, stretch fabric
- Easy-to-spot CAT 2/2112 label on the left sleeve leave no question that you're protected on the job site
- Hazard: Arc Flash, Flash Fire
- Wash Care: Industrial Laundry - Light Soil
- Fabric: Body: 6.75 oz. (230 g/m2) Flame Resistant Jersey
- Blend: Body: 93% Cotton/7% Spandex Elastane
- Country of Origin: Imported, Made in USA Fabric
Specifications
Specifications
Size | Small |
Gender | Men's |
Material | Cotton, Spandex |
Color | Navy |
Neck Style | Collar |
Sleeve Style | Long Sleeve |
Construction | Knit |
Closure Type | Button |
Disposable | Reusable |
No. of Pockets | 2 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction