Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bulwark™ Men's Relaxed Fit Bootcut Flame Resistant Jean with Stretch
Click to view available options
Size:
30 x 30 in.
30 x 32 in.
30 x 34 in.
32 x 30 in.
32 x 32 in.
32 x 34 in.
32 x 36 in.
33 x 30 in.
33 x 32 in.
33 x 34 in.
33 x 36 in.
34 x 30 in.
34 x 32 in.
34 x 34 in.
34 x 36 in.
35 x 30 in.
35 x 32 in.
35 x 34 in.
35 x 36 in.
36 x 30 in.
36 x 32 in.
36 x 34 in.
36 x 36 in.
38 x 30 in.
38 x 32 in.
38 x 34 in.
38 x 36 in.
40 x 30 in.
40 x 32 in.
40 x 34 in.
42 x 30 in.
42 x 32 in.
42 x 34 in.
42 x 36 in.
44 x 30 in.
44 x 32 in.
44 x 34 in.
44 x 36 in.
46 x 30 in.
46 x 32 in.
46 x 34 in.
48 x 30 in.
48 x 32 in.
48 x 34 in.
50 x 30 in.
50 x 32 in.
50 x 34 in.
Description
- Sits below natural waist with relaxed fit through waist, hip and thigh
- Bootcut leg opening fits easily over boots
- One-piece waistband with metal button closure (metal is not exposed to skin)
- Two hip pockets and scoop front pockets
- Larger belt loop and waistband
- Contrasting, two-toned stitching
- Stretch fabric inseam and seat seam
- Hazard: Flash Fire, Arc Flash
- Wash Care: Industrial Laundry - Light Soil, Home Wash
- Fabric: Flame-resistant, 12 oz. (405 g/m2)
- Blend: 94% Cotton/5% Polyester/1% Spandex
- Country of Origin: Imported, Made in USA Fabric
Specifications
Specifications
Size | 48 x 32 in. |
Gender | Men's |
Material | Cotton, Polyester, Spandex |
Color | Denim |
Seam Type | Stitched |
No. of Pockets | 4 |
Closure Type | Zipper |
Disposable | Reusable |
Hazard Risk Category | Category 2 |
Arc Flash Rating | 15 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction