Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bulwark™ Men's Straight Fit Bootcut Flame Resistant Jean with Stretch

Catalog No. 17204474
Click to view available options
Size:
30 x 30 in.
30 x 32 in.
30 x 34 in.
32 x 30 in.
32 x 32 in.
32 x 34 in.
32 x 36 in.
33 x 30 in.
33 x 32 in.
33 x 34 in.
33 x 36 in.
34 x 30 in.
34 x 32 in.
34 x 34 in.
34 x 36 in.
35 x 30 in.
35 x 32 in.
35 x 34 in.
35 x 36 in.
36 x 30 in.
36 x 32 in.
36 x 34 in.
36 x 36 in.
38 x 30 in.
38 x 32 in.
38 x 34 in.
38 x 36 in.
40 x 30 in.
40 x 32 in.
40 x 34 in.
40 x 36 in.
42 x 30 in.
42 x 32 in.
42 x 34 in.
42 x 36 in.
44 x 30 in.
44 x 32 in.
44 x 34 in.
44 x 36 in.
46 x 30 in.
46 x 32 in.
46 x 34 in.
48 x 32 in.
48 x 34 in.
50 x 30 in.
50 x 32 in.
50 x 34 in.

Men's Straight Fit Flame Resistant Jean with Stretch are made from 12 oz. flame-resistant 94% cotton, 5% polyester, 1% spandex denim that provides comfort and mobility while you work.

  • Straight fit with a larger waistband for comfort and durability
  • One-piece waistband with concealed, metal button closure
  • Two patch spade shaped hip pockets and scoop front pockets
  • Larger belt loop and waistband
  • Stretch fabric with felled inseam and seat seam
  • Hazard: Flash Fire, Arc Flash
  • Wash Care: Industrial Laundry - Light Soil, Home Wash
  • Fabric: Flame-resistant, 12 oz. (405 g/m2)
  • Blend: 94% Cotton/5% Polyester/1% Spandex
  • Country of Origin: Imported, Made in USA Fabric

Specifications

Size 35 x 36 in.
Gender Men's
Material Cotton, Polyester, Spandex
Color Denim
Seam Type Stitched
No. of Pockets 4
Closure Type Button
Disposable Reusable
Hazard Risk Category Category 2
Arc Flash Rating 15
Certifications/Compliance NFPA 2112
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.