Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mena Antibody (ENAH/1988), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279887H
Description
Mena Monoclonal specifically detects Mena in Human samples. It is validated for Western Blot, ELISA.Specifications
Mena | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
ENAH | |
Recombinant fragment of human Mena protein (around aa 485-589) (exact sequence is proprietary) (Uniprot: Q8N8S7) | |
0.1 mL | |
Cancer, Neuroscience | |
Human | |
IgG2c κ |
Western Blot, ELISA | |
ENAH/1988 | |
Western Blot, ELISA | |
ENA, enabled homolog (Drosophila), FLJ10773, MENANDPP1, protein enabled homolog | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction