Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Menin Antibody (8G11), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP315181
This item is not returnable.
View return policy
Description
Menin Monoclonal antibody specifically detects Menin in Human samples. It is validated for Western Blot, ELISASpecifications
Menin | |
Monoclonal | |
Unconjugated | |
PBS (pH 7.4), 150mM NaCl, and 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA | |
8G11 | |
Western Blot 1:500-1:5000, ELISA | |
menin, multiple, multiple endocrine neoplasia I | |
A synthesized peptide derived from human Menin. UniProt ID (O00255). | |
100 μg | |
Cancer, DNA Repair, Transcription Factors and Regulators, Tumor Suppressors | |
4221 | |
Store at -20 to -80°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction