Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Menin Antibody, DyLight 550, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB100287R
Description
Menin Polyclonal antibody specifically detects Menin in Human samples. It is validated for Western BlotSpecifications
Menin | |
Polyclonal | |
Western Blot | |
menin, multiple, multiple endocrine neoplasia I | |
The immunogen recognized by this antibody maps to a region between residue 575 and the C-terminus (residue 615) of human Multiple Endocrine Neoplasia 1 using the numbering given in entry NP_000235.2 (GeneID 4221). | |
0.1 mL | |
Cancer, DNA Repair, Transcription Factors and Regulators, Tumor Suppressors | |
4221 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
DyLight 550 | |
50mM Sodium Borate | |
Goat | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction