Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals Menin Antibody [PE], Novus Biologicals™
SDP

Catalog No. AF6005P Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF6005P 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AF6005P Supplier Novus Biologicals Supplier No. AF6005P
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Menin Polyclonal antibody specifically detects Menin in Human samples. It is validated for Western Blot

Specifications

Antigen Menin
Applications Western Blot
Classification Polyclonal
Conjugate PE
Formulation PBS
Gene Alias menin, multiple, multiple endocrine neoplasia I
Host Species Goat
Immunogen E. coli-derived recombinant human Menin, aa 555-615, Accession # O00255
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, DNA Repair, Transcription Factors and Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 4221
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.