Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Menin Antibody (SR2123), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP321766100UL
Description
Menin Monoclonal antibody specifically detects Menin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, ImmunoprecipitationSpecifications
Menin | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
SR2123 | |
Western Blot 1:1000-1:5000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
menin, multiple, multiple endocrine neoplasia I | |
A synthesized peptide derived from human Menin (Uniprot #: O00255) | |
100 μg | |
Cancer, DNA Repair, Transcription Factors and Regulators, Tumor Suppressors | |
4221 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction