Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Menin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120338
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120338 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120338 Supplier Invitrogen™ Supplier No. PA5120338
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HeLa, HepG2, K-562. The target is usually found in the following locations: Nucleus. Immunogen sequence: QKLLWLLYDL GHLERYPMAL GNLADLEELE PTPGRPDPLT LYHKGIASAK TYYRDEHIYP YMYLAGYHCR NRNVREALQA WADTATVIQD YNYCREDEEI YKEFFEVAND VIPNLLKEAA SLLEAGEERP GEQSQGTQSQ GSALQDPECF AHLLRFYDGI CKWEEGSPTP VLHVGWATFL VQSLGRFEGQ VRQKVRIVSR EAEAAEAEEP WGEEAREGRR RGPRRESKPE EPPPPKKPAL DKGLGTGQGA VSGPPRKPPG TVAGTARGPE GGSTAQVPAP AASPPPEGPV LTFQSEKMKG MKELLVATKI NSSAIKLQLT AQSQVQMKKQ KVSTPSDYTL SFLKRQRKGL.

Potassium channels are a group of ubiquitously expressed proteins that serve numerous functions in excitable and non-excitable cells. One class of integral membrane potassium channels is the large conductance, calcium-activated potassium channel (Maxi K+). Maxi K+ differs from most other potassium channels in that its activation is controlled by both increases in intracellular calcium and by membrane depolarization. Maxi K+ dual activation is possible because of its structure. The core of the channel, which is similar to other potassium channels, is a Maxi K+ alpha homotetramer that contains both a voltage sensor and an intracellular calcium binding domain. In vascular smooth muscle, an auxiliary beta-subunit is found in a 1:1 stoichiometry. The beta-subunit exhibits its effect on the Maxi K+ channel by effectively decreasing by 5- to 10- fold the concentration of calcium required to keep the pore open.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Menin
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.61 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene MEN1
Gene Accession No. O00255, O88559
Gene Alias AW045611; MEAI; MEN1; Menin; menin 1; multiple endocrine neoplasia 1; multiple endocrine neoplasia I; SCG2
Gene Symbols MEN1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 266-615 of human Menin (NP_000235.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17283, 4221
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.