Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Menin Rabbit anti-Human, Mouse, Rat, Clone: 5M7L9, Novus Biologicals™

Rabbit Monoclonal Antibody
$206.50 - $458.50
Specifications
| Antigen | Menin |
|---|---|
| Clone | 5M7L9 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
Description
Menin Monoclonal antibody specifically detects Menin in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| Menin | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, DNA Repair, Transcription Factors and Regulators, Tumor Suppressors | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 4221 | |
| IgG | |
| Affinity purified |
| 5M7L9 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| menin, multiple, multiple endocrine neoplasia I | |
| A synthetic peptide corresponding to a sequence within amino acids 516-615 of human Menin (O00255). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title