Learn More
Invitrogen™ Menin Recombinant Rabbit Monoclonal Antibody (5M7L9)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA542807
Description
Positive test controls include: HeLa, HCT116, HepG2, NIH/3T3, Mouse lung, Mouse brain, Rat lung, Rat brain. The target is usually found in the following locations: Nucleus. Immunogen sequence: VSGPPRKPPG TVAGTARGPE GGSTAQVPAP TASPPPEGPV LTFQSEKMKG MKELLVATKI NSSAIKLQLT AQSQVQMKKQ KVSTPSDYTL SFLKRQRKGL.
Potassium channels are a group of ubiquitously expressed proteins that serve numerous functions in excitable and non-excitable cells. One class of integral membrane potassium channels is the large conductance, calcium-activated potassium channel (Maxi K+). Maxi K+ differs from most other potassium channels in that its activation is controlled by both increases in intracellular calcium and by membrane depolarization. Maxi K+ dual activation is possible because of its structure. The core of the channel, which is similar to other potassium channels, is a Maxi K+ alpha homotetramer that contains both a voltage sensor and an intracellular calcium binding domain. In vascular smooth muscle, an auxiliary beta-subunit is found in a 1:1 stoichiometry. The beta-subunit exhibits its effect on the Maxi K+ channel by effectively decreasing by 5- to 10- fold the concentration of calcium required to keep the pore open.
Specifications
Menin | |
Recombinant Monoclonal | |
0.58 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O00255, O88559, Q9WVR8 | |
MEN1 | |
A synthetic peptide corresponding to a sequence within amino acids 516-615 of human Menin (O00255). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Western Blot | |
5M7L9 | |
Unconjugated | |
MEN1 | |
AW045611; MEAI; MEN1; Menin; menin 1; multiple endocrine neoplasia 1; multiple endocrine neoplasia I; SCG2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
17283, 29417, 4221 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.