Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bulwark™ Men's Charcoal Lightweight Long Sleeve Flame Resistant T-shirt
Click to view available options
Size:
2X-Large
2X-Large (Long)
3X-Large
3X-Large (Long)
4X-Large
4X-Large (Long)
Large
Large (Long)
Medium
Small
X-Large
X-Large (Long)
Description
- Underarm gusset for improved breathability
- Single spade chest pocket
- Rib-knit crewneck collar
- Hazard: Flash Fire, Arc Flash
- Wash Care: Home Wash
- Fabric: EXCEL FR™ Flame resistant, 7 oz. (235 g/m2) Interlock
- Blend: 100% Cotton
- Country of Origin: Imported, Made in USA Fabric
Specifications
Specifications
Size | Small |
Gender | Men's |
Material | FR Cotton |
Color | Charcoal |
Neck Style | Hemmed Crew |
Sleeve Style | Long Sleeve |
Disposable | Reusable |
No. of Pockets | 1 |
Hazard Risk Category | Category 2 |
Arc Flash Rating | 8.9 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction