Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MENT Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | C1orf56 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Agglutination |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
MENT Polyclonal specifically detects MENT in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Aggregation.Specifications
C1orf56 | |
Polyclonal | |
Rabbit | |
Human | |
chromosome 1 open reading frame 56, FLJ20519, hypothetical protein LOC54964, MENT, RP11-316M1.9 | |
C1ORF56 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Agglutination | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
54964 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:SSTRFIANSQEPEIRLTSSLPRSPGRSTEDLPGSQATLSQWSTPGSTPSRWPSPSPTAMPSPEDLRLVLMPWGPWHCHCKSGTMS | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title