Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MENT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598897
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598897 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598897 Supplier Invitrogen™ Supplier No. PA598897
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MENT Polyclonal Antibody for Western Blot, IHC (P), ELISA

C1orf56, also named as MENT, AF1Q and MLLT11, is involved in control of cellular proliferation. It is highly expressed in several cancers especially in neuroblastoma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MENT
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 3.37 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene C1orf56
Gene Accession No. Q5XI62, Q9BUN1
Gene Alias C1orf56; chromosome 1 open reading frame 56; Gm128; MENT; methylated in normal thymocytes protein; pmis1; predicted gene 128; protein MENT; protein missing in infertile spermatozoa 1; RGD1359334; similar to hypothetical protein FLJ20519; UNQ547/PRO1104
Gene Symbols C1orf56, RGD1359334
Host Species Rabbit
Immunogen Recombinant Human Protein MENT protein (24-197AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 295265, 54964
Target Species Human, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.