Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MEOX1 Monoclonal Antibody (OTI5A3), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA804738

Description
Voltage-gated proton (hydrogen) channels play an important role in cellular defense against acidic stress. They are unique among ion channels with respect to their extremely high selectivity, marked temperature dependence, and unitary conductance, which is 3 orders of magnitude lower than that of most other ion channels. NOX1 is a homolog of the catalytic subunit of the superoxide-generating NADPH oxidase of phagocytes, gp91phox. Two transcript variants encoding different isoforms have been found for this gene.Specifications
MEOX1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P50221 | |
MEOX1 | |
Full length human recombit protein of human MEOX1 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
OTI5A3 | |
Unconjugated | |
MEOX1 | |
KFS2, MOX1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
4222 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction