Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEOX1 Monoclonal Antibody (OTI5B11), TrueMAB™, OriGene
SDP

Catalog No. 501680378
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-0378 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-0378 Supplier OriGene Supplier No. TA804739
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Voltage-gated proton (hydrogen) channels play an important role in cellular defense against acidic stress. They are unique among ion channels with respect to their extremely high selectivity, marked temperature dependence, and unitary conductance, which is 3 orders of magnitude lower than that of most other ion channels. NOX1 is a homolog of the catalytic subunit of the superoxide-generating NADPH oxidase of phagocytes, gp91phox. Two transcript variants encoding different isoforms have been found for this gene.

Specifications

Antigen MEOX1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI5B11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene MEOX1
Gene Accession No. P50221
Gene Alias KFS2, MOX1
Gene Symbols MEOX1
Host Species Mouse
Immunogen Full length human recombit protein of human MEOX1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4222
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.