Learn More
Invitrogen™ MEOX2 Polyclonal Antibody

Description
Positive Control: Rat kidney tissue lysates, rat kidney tissue. Subcellular Location: Nucleus.
Specifications
Specifications
| Antigen | MEOX2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | MEOX2 |
| Gene Accession No. | P39020, P50222 |
| Gene Alias | AI528662; GAX; Growth arrest-specific homeobox; homeobox protein MOX-2; MEOX2; mesenchyme homeo box 2 (growth arrest-specific homeo box); mesenchyme homeobox 2; Mox2; Mox-2 |
| Gene Symbols | MEOX2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.