Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MEP1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528371
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528371 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528371 Supplier Invitrogen™ Supplier No. PA528371
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (80%), Rat (85%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

MEP1A (Meprin A Subunit Alpha) is a Protein Coding gene. Among its related pathways are Collagen chain trimerization. Gene Ontology (GO) annotations related to this gene include metalloendopeptidase activity and metallopeptidase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MEP1A
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene MEP1A
Gene Accession No. Q16819
Gene Alias AI098089; AW107200; bA268F1.1 (meprin A alpha (PABA peptide hydrolase)); E-24.18; Endopeptidase-2; endopeptidase-24.18 subunit alpha; Mep1; Mep-1; MEP1A; Mep-1a; meprin 1 alpha; meprin A alpha; meprin A alpha-subunit; meprin A subunit alpha; meprin A, alpha (PABA peptide hydrolase); meprin alpha; N-benzoyl-L-tyrosyl-P-amino-benzoic acid hydrolase subunit alpha; PABA peptide hydrolase; PPH alpha; PPHA
Gene Symbols MEP1A
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the Extracellular domain of human Meprin A alpha. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4224
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.