Learn More
Invitrogen™ MEP1A Polyclonal Antibody

Description
Immunogen sequence: MSSSMVFTTS KSHTSPAIND TVIWDRPSRV GTYHTDCNCF RSIDLGWSGF ISHQMLKRRS FLKNDDLIIF VDFEDITHLS QTEVPTKGKR LSPQGLILQG QEQQVSEEGS GKAMLEEALP VSLSQGQPSR QKRSVE Highest antigen sequence indentity to the following orthologs: Rat - 63%, Mouse - 65%.
Specifications
Specifications
| Antigen | MEP1A |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | MEP1A |
| Gene Accession No. | Q16819 |
| Gene Alias | AI098089; AW107200; bA268F1.1 (meprin A alpha (PABA peptide hydrolase)); E-24.18; Endopeptidase-2; endopeptidase-24.18 subunit alpha; Mep1; Mep-1; MEP1A; Mep-1a; meprin 1 alpha; meprin A alpha; meprin A alpha-subunit; meprin A subunit alpha; meprin A, alpha (PABA peptide hydrolase); meprin alpha; N-benzoyl-L-tyrosyl-P-amino-benzoic acid hydrolase subunit alpha; PABA peptide hydrolase; PPH alpha; PPHA |
| Gene Symbols | MEP1A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.