Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MEP1A Recombinant Rabbit Monoclonal Antibody (009), Invitrogen™
GREENER_CHOICE

Catalog No. PIMA543125
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543125 100 μL
PIMA543435 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543125 Supplier Invitrogen™ Supplier No. MA543125
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Specificity: Human MEP1A

MEP1A (Meprin A Subunit Alpha) is a Protein Coding gene. Among its related pathways are Collagen chain trimerization. Gene Ontology (GO) annotations related to this gene include metalloendopeptidase activity and metallopeptidase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MEP1A
Applications ELISA
Classification Recombinant Monoclonal
Clone 009
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene MEP1A
Gene Accession No. Q16819
Gene Alias AI098089; AW107200; bA268F1.1 (meprin A alpha (PABA peptide hydrolase)); E-24.18; Endopeptidase-2; endopeptidase-24.18 subunit alpha; Mep1; Mep-1; MEP1A; Mep-1a; meprin 1 alpha; meprin A alpha; meprin A alpha-subunit; meprin A subunit alpha; meprin A, alpha (PABA peptide hydrolase); meprin alpha; N-benzoyl-L-tyrosyl-P-amino-benzoic acid hydrolase subunit alpha; PABA peptide hydrolase; PPH alpha; PPHA
Gene Symbols MEP1A
Host Species Rabbit
Immunogen Recombinant Human MEP1A protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4224
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.