Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MEP1B Monoclonal Antibody (09)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA530668
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human MEP1B/Meprin beta.
Meprins are multidomain zinc metalloproteases that are highly expressed in mammalian kidney and intestinal brush border membranes, and in leukocytes and certain cancer cells. They are involved in the hydrolysis of a variety of peptide and protein substrates, and have been implicated in cancer and intestinal inflammation. Mature meprins are oligomers of evolutionarily related, but separately encoded alpha and/or beta subunits. Homooligomers of alpha subunit are secreted, whereas, oligomers containing the beta subunit are plasma membrane-bound. This gene encodes the beta subunit. Targeted disruption of this gene in mice affects embryonic viability, renal gene expression profiles, and distribution of the membrane-associated alpha subunit in kidney and intestine.
Specifications
MEP1B | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7 | |
Q16820 | |
Mep1b | |
Recombinant Human MEP1B/Meprin beta Protein (Met1-Thr652). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA | |
09 | |
Unconjugated | |
Mep1b | |
EC 3.4.24.63; Endopeptidase-2; MEP1B; Mep-1b; meprin 1 beta; meprin A; meprin A subunit beta; meprin A, beta; meprin B; meprin beta; N-benzoyl-L-tyrosyl-p-amino-benzoic acid hydrolase beta subunit; N-benzoyl-L-tyrosyl-P-amino-benzoic acid hydrolase subunit beta; PABA peptide hydrolase; PPH beta | |
Mouse | |
Protein A | |
RUO | |
4225 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction