Learn More
MEPCE Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | MEPCE |
---|---|
Applications | Immunohistochemistry, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-7074
![]() |
Bethyl Laboratories, Inc
A304184AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
S-adenosyl-L-methionine-dependent methyltransferase that adds a methylphosphate cap at the 5′-end of 7SK snRNA, leading to stabilize it.Specifications
MEPCE | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
MEPCE; methylphosphate capping enzyme; BCDIN3; FLJ20257; bin3, bicoid-interacting 3; bin3, bicoid-interacting 3, homolog | |
Between 200 and 250 | |
Primary | |
4° C | |
MEPCE |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q7L2J0, Q8K3A9 | |
231803, 56257 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.