Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MEPE/OF45 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP316826100UL
This item is not returnable.
View return policy
Description
MEPE/OF45 Polyclonal antibody specifically detects MEPE/OF45 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
MEPE/OF45 | |
Polyclonal | |
PBS, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
Unconjugated | |
extracellular phosphoglycoprotein with ASARM motif (bone), matrix extracellular phosphoglycoprotein, OF45 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 90-256 of human MEPE/OF45 (NP_064588.1). | |
100 μg | |
Cell Biology, Cytoskeleton Markers, Signal Transduction | |
56955 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction