Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Meprin alpha Subunit/MEP1A Antibody (001), mFluor Violet 450 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290383MFV450
Description
Meprin alpha Subunit/MEP1A Monoclonal antibody specifically detects Meprin alpha Subunit/MEP1A in Mouse samples. It is validated for ELISA, Immunohistochemistry (Paraffin)Specifications
Meprin alpha Subunit/MEP1A | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
ELISA, Immunohistochemistry (Paraffin) | |
001 | |
ELISA, Immunohistochemistry-Paraffin | |
bA268F1.1 (meprin A alpha (PABA peptide hydrolase)), EC 3.4.24, EC 3.4.24.18, endopeptidase-2, meprin A subunit alpha, meprin A, alpha (PABA peptide hydrolase), N-benzoyl-L-tyrosyl-P-amino-benzoic acid hydrolase subunit alpha, PABA peptide hydrolase, PPH alpha, PPHA | |
Rabbit immunized with purified, recombinant Mouse Meprin alpha Subunit/MEP1A (Accession#: NP_032611.2, Met 1-Arg 615) | |
0.1 mL | |
Proteases & Other Enzymes | |
4224 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction