Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Meprin alpha Subunit/MEP1A Antibody (364312), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB3220N
Description
Meprin alpha Subunit/MEP1A Monoclonal antibody specifically detects Meprin alpha Subunit/MEP1A in Human samples. It is validated for Western Blot, Immunoprecipitation, Flow Cytometry, CyTOFSpecifications
Meprin alpha Subunit/MEP1A | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunoprecipitation, Flow Cytometry, CyTOF | |
364312 | |
Western Blot, Immunoprecipitation, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
bA268F1.1 (meprin A alpha (PABA peptide hydrolase)), EC 3.4.24, EC 3.4.24.18, endopeptidase-2, meprin A subunit alpha, meprin A, alpha (PABA peptide hydrolase), N-benzoyl-L-tyrosyl-P-amino-benzoic acid hydrolase subunit alpha, PABA peptide hydrolase, PPH alpha, PPHA | |
Mouse myeloma cell line NS0-derived recombinant human Meprin alpha Subunit/MEP1A, Val22-Gln601, Accession # AAA21338 | |
0.1 mL | |
Proteases & Other Enzymes | |
4224 | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction