Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Meprin beta Subunit/MEP1B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$231.00 - $262.00
Specifications
Antigen | Meprin beta Subunit/MEP1B |
---|---|
Dilution | ELISA 1:5000-1:10000 |
Applications | ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Meprin beta Subunit/MEP1B Polyclonal antibody specifically detects Meprin beta Subunit/MEP1B in Human samples. It is validated for ELISASpecifications
Meprin beta Subunit/MEP1B | |
ELISA | |
Unconjugated | |
Rabbit | |
Proteases & Other Enzymes | |
0.2 um filtered solution in PBS | |
4225 | |
IgG | |
Antigen and protein A Affinity-purified |
ELISA 1:5000-1:10000 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
EC 3.4.24.63, endopeptidase-2, meprin A subunit beta, meprin A, beta, Meprin B, N-benzoyl-L-tyrosyl-p-amino-benzoic acid hydrolase beta subunit, N-benzoyl-L-tyrosyl-P-amino-benzoic acid hydrolase subunit beta, PABA peptide hydrolase, PPH beta | |
Produced in rabbits immunized with purified, recombinant Human Meprin beta Subunit/MEP1B (Accession#: NP_005916.2; Met1-Thr652) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title