Learn More
Thermo Scientific™ MEPS™ for GC: CTC Analytics Using 250¦L Syringes
Save hours in sample preparation, and go from extraction to injection in a single process with these 250μL syringes for GC applications using CTC Analytics.
Specifications
For Use With (Application) | Gas Chromatography |
---|---|
For Use With (Equipment) | CTC Analytics Systems Using 250 μL Syringes |
Product Line | HyperSep |
Catalog Number | Mfr. No. | Description | Sorbent | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Sorbent | Price | Quantity & Availability | ||||
60-308-301
|
Thermo Scientific™
60308301 |
Retain PEP MEPS for GC; For CTC | HyperSep Retain PEP MEPS | N/A | N/A | N/A | |||
60-308-302
|
Thermo Scientific™
60308302 |
Retain-CX MEPS for GC; For CTC | HyperSep Retain CX MEPS | N/A | N/A | N/A | |||
60-308-303
|
Thermo Scientific™
60308303 |
Retain-AX MEPS for GC; For CTC | HyperSep Retain AX MEPS | N/A | N/A | N/A | |||
60-308-304
|
Thermo Scientific™
60308304 |
Hypercarb MEPS for GC; For CTC | HyperSep Hypercarb MEPS | N/A | N/A | N/A | |||
60-308-305
|
Thermo Scientific™
60308305 |
Verify-CX MEPS for GC; For CTC | HyperSep Verify CX MEPS | N/A | N/A | N/A | |||
60-308-306
|
Thermo Scientific™
60308306 |
Verify-AX MEPS for GC; For CTC | HyperSep Verify AX MEPS | N/A | N/A | N/A | |||
60-308-307
|
Thermo Scientific™
60308307 |
C18 MEPS for GC; For CTC | HyperSep C18 MEPS | N/A | N/A | N/A | |||
60-308-308
|
Thermo Scientific™
60308308 |
Silica MEPS for GC; For CTC | HyperSep Silica MEPS | N/A | N/A | N/A | |||
60-308-309
|
Thermo Scientific™
60308309 |
MEPS Development Kit for GC | HyperSep Retain PEP, Retain CX, Retain AX, Hypercarb, and C18 | N/A | N/A | N/A | |||
Description
Save hours in sample preparation, and go from extraction to injection in a single process with Thermo Scientific™ HyperSep™ MEPS™ (Micro Extraction by Packed Sorbent) technology.
Suitable for reversed phases, normal phases, mixed mode or ion exchange chemistries, Thermo Scientific HyperSep MEPS Syringes and Components for Analytics are available in a variety of SPE phase chemistries. The HyperSep MEPS Barrel Insert and Needle Assembly (BIN) contains the stationary phase, and is built into the syringe needle.
These HyperSep MEPS BINs are designed for use with Thermo Scientific, CTC Analytics, HTA and Varian 8400 GC Systems.
MEPS performs the same functions as SPE — the removal of interfering matrix components and the selective isolation and concentration of analytes — except that it miniaturizes conventional SPE packed bed devices from milliliter bed volumes to microliter volumes, and incorporates them right into the syringe.Mechanically robust, the technique offers significant advantages over conventional SPE extraction for method development and other GC applications:
- Significantly reduces the time to prepare and inject samples from hours to minutes — typically less than two minutes per sample
- Eliminates all extra steps between sample preparation and sample injection; the technique is fully compatible with direct injection onto GC
- Reduces buffer and solvent volume from milliliters to microliters
- Reduces the required sample volume to as little as 3.6μL, making the techniques especially well-suited for on-line use with volume-limited samples
- Can easily be automated compatible and is compatible with CTC and eVol platforms
Specifications
Gas Chromatography | |
CTC Analytics Systems Using 250 μL Syringes | |
HyperSep |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.