Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mer Antibody (108928) [Janelia Fluor™ 646], Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB5912J
Description
Mer Monoclonal antibody specifically detects Mer in Mouse samples. It is validated for Flow Cytometry, CyTOFSpecifications
Mer | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry, CyTOF | |
108928 | |
Flow Cytometry, CyTOF-ready | |
c-mer, c-mer proto-oncogene tyrosine kinase, EC 2.7.10, EC 2.7.10.1, MER, MER receptor tyrosine kinase, MGC133349, Receptor tyrosine kinase MerTK, RP38Proto-oncogene c-Mer, STK kinase, tyrosine-protein kinase Mer | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse Mer, Glu23-Phe498, Accession # Q60805 | |
0.1 mL | |
Cancer, Protein Kinase, Signal Transduction, Vision | |
10461 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction