Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mer Antibody (108928), mFluor Violet 610 SE, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB5912MFV610
Description
Mer Monoclonal antibody specifically detects Mer in Mouse samples. It is validated for Flow Cytometry, CyTOFSpecifications
| Mer | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rat | |
| 0.1 mL | |
| Cancer, Phospho Specific, Protein Kinase, Signal Transduction, Vision | |
| 10461 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF | |
| 108928 | |
| Flow Cytometry, CyTOF-ready | |
| c-mer, c-mer proto-oncogene tyrosine kinase, EC 2.7.10, EC 2.7.10.1, MER, MER receptor tyrosine kinase, MGC133349, Receptor tyrosine kinase MerTK, RP38Proto-oncogene c-Mer, STK kinase, tyrosine-protein kinase Mer | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse Mer, Glu23-Phe498, Accession # Q60805 | |
| RUO | |
| Primary | |
| Mouse | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction