Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mer Antibody (8H11), Novus Biologicals™

Human Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Mer |
---|---|
Clone | 8H11 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Description
Mer Monoclonal antibody specifically detects Mer in Human samples. It is validated for ELISASpecifications
Mer | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
c-mer, c-mer proto-oncogene tyrosine kinase, EC 2.7.10, EC 2.7.10.1, MER, MER receptor tyrosine kinase, MGC133349, Receptor tyrosine kinase MerTK, RP38Proto-oncogene c-Mer, STK kinase, tyrosine-protein kinase Mer | |
Recombinant Human Mer protein [UniProt Q12866] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
8H11 | |
ELISA | |
Unconjugated | |
Human | |
Cancer, Phospho Specific, Protein Kinase, Signal Transduction, Vision | |
50% Glycerol, 0.01M PBS, PH 7.4 | |
10461 | |
IgG4 | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title