Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mer Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1002309PE
Description
Mer Polyclonal antibody specifically detects Mer in Human samples. It is validated for Western BlotSpecifications
Mer | |
Polyclonal | |
Western Blot | |
c-mer, c-mer proto-oncogene tyrosine kinase, EC 2.7.10, EC 2.7.10.1, MER, MER receptor tyrosine kinase, MGC133349, Receptor tyrosine kinase MerTK, RP38Proto-oncogene c-Mer, STK kinase, tyrosine-protein kinase Mer | |
Maps to a region between residues 200 and 250 of human c-mer proto-oncogene tyrosine kinase using the numbering given in SwissProt entry Q12866 (GeneID 10461). | |
0.1 mL | |
Cancer, Phospho Specific, Protein Kinase, Signal Transduction, Vision | |
10461 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
PE | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction