Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mer Rat anti-Mouse, Brilliant Violet 605, Clone: 108928, BD OptiBuild
Rat Monoclonal Antibody
Manufacturer: BD Biosciences 747894
Description
Mer Monoclonal antibody specifically detects Mer in Mouse samples. It is validated for Flow Cytometry.Specifications
Mer | |
108928 | |
Brilliant Violet 605 | |
Rat | |
50 μg | |
Refrigerate (2 to 8°C) | |
Monoclonal | |
Mouse |
Flow Cytometry | |
0.2 mg/mL | |
Conjugated, Purified | |
IgG2a | |
RUO | |
Primary | |
Mer |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title