Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mer2, p Ser30 Antibody, Novus Biologicals™
SDP

Catalog No. NBP177754 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP177754 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP177754 Supplier Novus Biologicals Supplier No. NBP177754
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Mer2 Polyclonal specifically detects Mer2 in Yeast samples. It is validated for Western Blot, ELISA, Immunoprecipitation.

Specifications

Antigen Mer2 (p Ser30)
Applications Western Blot, ELISA, Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:10000, ELISA 1:5000-1:25000, Immunoprecipitation 1:100
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Accession No. P21651
Gene Alias MER2, REC107 Rec107p
Gene Symbols REC107
Host Species Rabbit
Immunogen This affinity purified Mer2 [p Ser30] Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to amino acids 26-35 of Saccharomyces cerevisiae Mer2 protein. (Uniprot: P21651)
Molecular Weight of Antigen 35.68 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 853478
Test Specificity This affinity-purified antibody is directed against the phosphorylated form of Saccharomyces cerevisiae Mer2 protein at the pS30 residue. The resultant affinity purified antibody was then cross-adsorbed against the non-phosphorylated peptide to remove any unwanted reactivities.
Target Species Yeast
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.