Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mer2Rabbit anti-Yeast, Polyclonal, Rockland™
SDP

Catalog No. 501991869
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1869 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-1869 Supplier Rockland Immunochemicals Supplier No. 600401925
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This affinity-purified antibody is directed against the Saccharomyces cerevisiae Mer2 protein. Reactivity occurs against Saccharomyces cerevisiae Mer2 protein and reactivity is independent of phosphorylation at residue S30. A BLAST analysis was used to suggest minimal cross reactivity with Mer2 homologues from other sources.

Mer2 (also known as meiotic recombination 2 protein) is a chromosomal protein that is critical for meiotic recombination and progression. It is phosphorylated at two serine residues, S30 and S271 by the yeast Cdk1 cyclin- dependent kinase homolog. This phosphorylation is S-phase specific, and thus has the potential to be a specific assay for S-phase cyclin-dependent kinases. Moreover, there are hints that the phosphorylation may be a mark of replication fork passage, which would indicate that S-phase CDK associates with the replication fork.

Specifications

Antigen Mer2
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.75 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Host Species Rabbit
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Yeast
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.