Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercuric Acetate, Powder, ACS, 98%, Spectrum™ Chemical
SDP

Catalog No. 18611529
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
25 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail

Chemical Identifiers

CAS 1600-27-7
Molecular Formula C4H6HgO4
Molecular Weight (g/mol) 318.68
InChI Key BRMYZIKAHFEUFJ-UHFFFAOYSA-L
IUPAC Name mercury(2+) diacetate
SMILES [Hg++].CC([O-])=O.CC([O-])=O

Specifications

CAS Max % 1
Quantity 2.5 kg
Percent Purity 98%
Grade ACS
Packaging Amber Glass Bottle

Caution: for manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.