Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercuric Acetate TS, 6% (w/v) in Glacial Acetic Acid, Ricca Chemical
SDP

Catalog No. p-3251906 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Amber Glass Bottle
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

Chemical Identifiers

CAS 1600-27-7, 64-19-7
Molecular Formula C4H6HgO4
Molecular Weight (g/mol) 318.68
InChI Key BRMYZIKAHFEUFJ-UHFFFAOYSA-L
Synonym mercury 2+ ion acetic acid
PubChem CID 15337
ChEBI CHEBI:33211
IUPAC Name mercury(2+);diacetate
SMILES CC(=O)[O-].CC(=O)[O-].[Hg+2]

Specifications

Color Colorless
CAS Min % 93.0, 5.3
CAS Max % 96.0, 5.52
Percent Purity 6%
Grade Laboratory
Packaging Amber Glass Bottle
Solvent or Matrix Glacial Acetic Acid
pH <2
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 6% (w/v)
Chemical Name or Material Mercuric Acetate, Test Solution
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.